Skip to Content
Merck
Ropivacaine

Ropivacaine

Ropivacaine
Synonym(s):
(2S)-N-(2,6-Dimethylphenyl)-1-propyl-2-piperidinecarboxamide, Ropivacaine
Empirical Formula (Hill Notation):
C17H26N2O
CAS Number:
Molecular Weight:
274.40

Products

Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing