Rivastigmine tartrate
Synonym(s):
ENA-713, Ethylmethyl-carbamic acid 3-[(1S)-1-(dimethylamino)ethyl]phenyl ester, N-Ethyl-N-methyl-carbamic acid 3-[(1S)-1-(dimethylamino)ethyl]phenyl ester tartrate, S-Rivastigmine tartrate
Empirical Formula (Hill Notation):
C14H22N2O2 · C4H6O6
CAS Number:
Molecular Weight:
400.42
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing