Skip to Content
Merck
vibrant-m

Rivastigmine for system suitability

vibrant-m
Synonym(s):
Rivastigmine tartrate, ENA-713, Ethylmethyl-carbamic acid 3-[(1S)-1-(dimethylamino)ethyl]phenyl ester, N-Ethyl-N-methyl-carbamic acid 3-[(1S)-1-(dimethylamino)ethyl]phenyl ester tartrate, S-Rivastigmine tartrate
Empirical Formula (Hill Notation):
C14H22N2O2 · C4H6O6
CAS Number:
Molecular Weight:
400.42

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1