Reichardt’s dye
Synonym(s):
2,6-Diphenyl-4-(2,4,6-triphenyl-1-pyridinio)phenolate, 2,6-Diphenyl-4-(2,4,6-triphenylpyridinio)phenolate
Empirical Formula (Hill Notation):
C41H29NO
CAS Number:
Molecular Weight:
551.68
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing