Skip to Content
Merck
Quinalizarin ≥95% (HPLC)

Quinalizarin

Quinalizarin ≥95% (HPLC)
Synonym(s):
1,2,5,8-Tetrahydroxy-9,10-anthraquinone, 1,2,5,8-Tetrahydroxyanthraquinone, Alizarin Bordeaux BD, Alizarinbordeaux, Alizarine Bordeaux, Alizarine Bordeaux B, C.I. 58500, C.I. Mordant Violet 26, Khinalizarin, NSC 144046, NSC 4896, PHF 016
Empirical Formula (Hill Notation):
C14H8O6
CAS Number:
Molecular Weight:
272.21
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1