Skip to Content
Merck
Pyribencarb PESTANAL®, analytical standard

Pyribencarb

Pyribencarb PESTANAL®, analytical standard
Synonym(s):
Methyl {2-chloro-5-[(1E)-1-(6-methyl-2-pyridylmethoxyimino)ethyl]benzyl}carbamate
Empirical Formula (Hill Notation):
C18H20ClN3O3
CAS Number:
Molecular Weight:
361.82

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique