![Propionaldehyde-2,4-DNPH certified reference material, vial of 100 mg, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/421/804/ff1aa2bd-2a20-4509-927f-c9eb1943ec12/640/ff1aa2bd-2a20-4509-927f-c9eb1943ec12.png)
Propionaldehyde-2,4-DNPH
![Propionaldehyde-2,4-DNPH certified reference material, vial of 100 mg, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/421/804/ff1aa2bd-2a20-4509-927f-c9eb1943ec12/640/ff1aa2bd-2a20-4509-927f-c9eb1943ec12.png)
Synonym(s):
Propionaldehyde-2,4-dinitrophenylhydrazone
Linear Formula:
C2H5CH=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
238.20
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing