Skip to Content
Merck
Potassium hydrogen phthalate NIST® SRM® 185i, pH standard

Potassium hydrogen phthalate

Potassium hydrogen phthalate NIST® SRM® 185i, pH standard
Synonym(s):
Potassium hydrogen phthalate, Potassium phthalate monobasic, KHP, Phthalic acid monopotassium salt, Potassium biphthalate
Linear Formula:
HOOCC6H4COOK
CAS Number:
Molecular Weight:
204.22

Products

Product Category
Brand
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique