Skip to Content
Merck
Poly(2-ethyl-2-oxazoline) average Mw ~500,000, PDI 3‑4

Poly(2-ethyl-2-oxazoline)

Poly(2-ethyl-2-oxazoline) average Mw ~500,000, PDI 3‑4
Synonym(s):
PEOX, POx, PetOx, poly 2-ethyloxazoline, polyethyloxazoline
Linear Formula:
[-N(COC2H5)CH2CH2-]n
CAS Number:

Products

Physical Form
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing