Skip to Content
Merck
Platycodin D phyproof® Reference Substance

Platycodin D

Platycodin D phyproof® Reference Substance
Synonym(s):
3β-(β-D-glucopyranosyloxy)-2β,16α,23,24-tetrahydroxy-O-D-apio-β-D-furanosyl-(1→3)-O-β-D-xylopyranosyl-(1→4)-O-6-deoxy-olean-12-en-28-oic acid
Empirical Formula (Hill Notation):
C57H92O28
CAS Number:
Molecular Weight:
1225.32

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3