Pilocarpine nitrate
Synonym(s):
Pilocarpine nitrate salt
Empirical Formula (Hill Notation):
C11H16N2O2 · HNO3
CAS Number:
Molecular Weight:
271.27
Products
Brand
Feature
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing