![Phosphated Riboflavin (Riboflavin Sodium Phosphate) Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/386/767/7a75b0e2-112c-47b9-9122-022dd54c9f15/640/7a75b0e2-112c-47b9-9122-022dd54c9f15.png)
Phosphated Riboflavin (Riboflavin Sodium Phosphate)
![Phosphated Riboflavin (Riboflavin Sodium Phosphate) Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/386/767/7a75b0e2-112c-47b9-9122-022dd54c9f15/640/7a75b0e2-112c-47b9-9122-022dd54c9f15.png)
Synonym(s):
Riboflavin 5′-monophosphate sodium salt hydrate
Empirical Formula (Hill Notation):
C17H20N4O9P · xNa+ · yH2O
Molecular Weight:
455.34 (anhydrous free acid basis)
EC Number:
Products
Brand
Formula Weight
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing