
Perfluoro-3-methoxypropanoic acid

Synonym(s):
2,2,3,3-Tetrafluoro-3-(trifluoromethoxy)propanoic acid
Empirical Formula (Hill Notation):
C4HF7O3
CAS Number:
Molecular Weight:
230.04
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing