Skip to Content
Merck
Peonidin 3-O-glucoside chloride ≥95% (HPLC)

Peonidin 3-O-glucoside chloride

Peonidin 3-O-glucoside chloride ≥95% (HPLC)
Synonym(s):
3-(Glucosyloxy)-4′,5,7-trihydroxy-3′-methoxyflavylium chloride, Glucopeonidin chloride
Empirical Formula (Hill Notation):
C22H23ClO11
CAS Number:
Molecular Weight:
498.86

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Research Area
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing