Skip to Content
Merck
Pentetic Acid Pharmaceutical Secondary Standard; Certified Reference Material

Pentetic acid

Pentetic Acid Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Diethylenetriaminepentaacetic acid, (Carboxymethylimino)bis(ethylenenitrilo)tetraacetic acid, N,N-Bis(2-[bis(carboxymethyl)amino]ethyl)glycine, DETAPAC, DTPA, Penta(carboxymethyl)diethylenetriamine, Pentetic acid
Linear Formula:
[(HOOCCH2)2NCH2CH2]2NCH2COOH
CAS Number:
Molecular Weight:
393.35

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade