
Penicillamine disulfide

Synonym(s):
D-Penicillamine disulfide, 3,3′-Dithiobis(2-amino-3-methylbutanoic acid), 3,3′-Dithiodi-D-valine, S,S′-Bi(D-penicillamine)
Empirical Formula (Hill Notation):
C10H20N2O4S2
CAS Number:
Molecular Weight:
296.41
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-3 of 3