Pefloxacin mesylate dihydrate
Synonym(s):
3-Quinolinecarboxylic acid, 1-ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-, monomethanesulfonate, dihydrate, Pefloxacine monomethanesulfonate dihydrate, Pefloxacinium methanesulfonate dihydrate
Linear Formula:
C17H20FN3O3 • CH4O3S • 2H2O
Products
Product Category
Brand
Formula Weight
Physical Form
Activity Spectrum
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing