PD 98059
Synonym(s):
PD 98059, 2-(2-Amino-3-methoxyphenyl)-4H-1-benzopyran-4-one, PD98059, InSolution PD 98059
Empirical Formula (Hill Notation):
C16H13NO3
CAS Number:
Molecular Weight:
267.28
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Research Area
Quality Segment
Showing 1-3 of 3