Skip to Content
Merck
Parathyroid Hormone Fragment 1-34 human ≥95% (HPLC), powder

Parathyroid Hormone Fragment 1-34 human

Parathyroid Hormone Fragment 1-34 human ≥95% (HPLC), powder
Synonym(s):
PTH 1-34, Parathormone (1-34)
Empirical Formula (Hill Notation):
C181H291N55O51S2
CAS Number:
Molecular Weight:
4117.72

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing