Paraformaldehyde
Synonym(s):
Polyoxymethylene
Linear Formula:
HO(CH2O)nH
CAS Number:
Molecular Weight:
30.03 (as monomer)
Products
Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Special Grade
Technique
Showing 1-4 of 4