![Panaxatriol phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/126/690/71b4c1d3-7d35-40d6-aa3f-73fb1180ea1d/640/71b4c1d3-7d35-40d6-aa3f-73fb1180ea1d.png)
Panaxatriol
![Panaxatriol phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/126/690/71b4c1d3-7d35-40d6-aa3f-73fb1180ea1d/640/71b4c1d3-7d35-40d6-aa3f-73fb1180ea1d.png)
Synonym(s):
(3β,6β,12β,20R)-20,25-epoxydammarane-3,6,12-triol, (3S,5R,6R,8R,9R,10R,12R,13R,14R,17S)-4,4,8,10,14-pentamethyl-17-[(2R)-2,6,6-trimethyloxan-2-yl]-2,3,5,6,7,9,11,12,13,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthrene-3,6,12-triol
Empirical Formula (Hill Notation):
C30H52O4
CAS Number:
Molecular Weight:
476.73
Products
Formula Weight
Manufacturer
Product Line
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing