Ophiopogonin D
Synonym(s):
(1β,3β,25R)-3-Hydroxyspirost-5-en-1-yl O-6-deoxy-α-L-mannopyranosyl-(1→2)-O-[β-D-xylopyranosyl-(1→3)]-6-deoxy-β-D-galactopyranoside, 25(R)-Ruscogenin 1-O-[α-L-rhamnopyranosyl-(1→2)][β-D-xylopyranosyl-(1→3)]-β-D-fucopyranoside, Deacetylophiopogonin C, OJV-V
Empirical Formula (Hill Notation):
C44H70O16
CAS Number:
Molecular Weight:
855.02
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing