Norgestrel
Synonym(s):
(±)-Norgestrel, (17α)-(±)-13-Ethyl-17-hydroxy-18,19-dinorpregn-4-en-20-yn-3-one
Empirical Formula (Hill Notation):
C21H28O2
CAS Number:
Molecular Weight:
312.45
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing