Skip to Content
Merck
Norflurazon PESTANAL®, analytical standard

Norflurazon

Norflurazon PESTANAL®, analytical standard
Synonym(s):
4-Chloro-5-(methylamino)-2-(α,α,α-trifluoro-m-tolyl)-3(2H)-pyridazinone
Empirical Formula (Hill Notation):
C12H9ClF3N3O
CAS Number:
Molecular Weight:
303.67
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing