Skip to Content
Merck
vibrant-m

Norfloxacin Related Compound K

vibrant-m
Synonym(s):
6-Fluoro-1-methyl-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid
Empirical Formula (Hill Notation):
C15H16FN3O3
CAS Number:
Molecular Weight:
305.30

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1