![Norfloxacin Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/311/070/feaa4085-1e4e-4812-abab-b22ab35365b1/640/feaa4085-1e4e-4812-abab-b22ab35365b1.jpg)
Norfloxacin Related Compound A
![Norfloxacin Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/311/070/feaa4085-1e4e-4812-abab-b22ab35365b1/640/feaa4085-1e4e-4812-abab-b22ab35365b1.jpg)
Synonym(s):
7-Chloro-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Empirical Formula (Hill Notation):
C12H9ClFNO3
CAS Number:
Molecular Weight:
269.66
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2