Skip to Content
Merck
N,N-Dimethylsphingosine ≥98% (HPLC)

N,N-Dimethylsphingosine

N,N-Dimethylsphingosine ≥98% (HPLC)
Synonym(s):
N,N-Dimethylsphingosine, (2S,3R,4E)-2-(Dimethylamino)-4-octadecene-1,3-diol, D-Erythro-N,N-Dimethylsphingosine, DMS, Dimethylsphingosine
Empirical Formula (Hill Notation):
C20H41NO2
CAS Number:
Molecular Weight:
327.55

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing