Skip to Content
Merck
N,N-Dimethylformamide-d7 ≥99.5 atom % D, contains 0.03 % (v/v) TMS

N,N-Dimethylformamide-d7

N,N-Dimethylformamide-d7 ≥99.5 atom % D, contains 0.03 % (v/v) TMS
Synonym(s):
DMF-d7, Heptadeutero-N,N-dimethylformamide
Linear Formula:
DCON(CD3)2
CAS Number:
Molecular Weight:
80.14

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Technique
Showing 1-2 of 2