![N-Methyl-perfluorooctane-1-sulfonamide certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/208/994/53fa7730-7001-4d42-9754-62fd03f0f50b/640/53fa7730-7001-4d42-9754-62fd03f0f50b.png)
N-Methyl-perfluorooctane-1-sulfonamide
![N-Methyl-perfluorooctane-1-sulfonamide certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/208/994/53fa7730-7001-4d42-9754-62fd03f0f50b/640/53fa7730-7001-4d42-9754-62fd03f0f50b.png)
Synonym(s):
1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluoro-N-methyl-1-octanesulfonamide, Perfluoro-N-methyloctanesulfonamide
Empirical Formula (Hill Notation):
C9H4F17NO2S
CAS Number:
Molecular Weight:
513.17
Products
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing