
Niflumic acid

Synonym(s):
2-(α,α,α-Trifluoro-m-toluidino)nicotinic acid, 2-[3-(Trifluoromethyl)anilino]nicotinic acid
Empirical Formula (Hill Notation):
C13H9F3N2O2
CAS Number:
Molecular Weight:
282.22
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing