![Nickel(II) nitrate hexahydrate puriss. p.a., ≥98.5% (KT)](/deepweb/assets/sigmaaldrich/product/structures/117/960/599b7698-bbe9-4e0c-abac-e3ddac743e75/640/599b7698-bbe9-4e0c-abac-e3ddac743e75.png)
Nickel(II) nitrate hexahydrate
![Nickel(II) nitrate hexahydrate puriss. p.a., ≥98.5% (KT)](/deepweb/assets/sigmaaldrich/product/structures/117/960/599b7698-bbe9-4e0c-abac-e3ddac743e75/640/599b7698-bbe9-4e0c-abac-e3ddac743e75.png)
Synonym(s):
Nickel(II) nitrate hexahydrate
Linear Formula:
Ni(NO3)2 · 6H2O
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Reagent Type
Core
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing