Skip to Content
Merck
vibrant-m

(NE)-N-[(1-methylpyridin-1-ium-2-yl)methylidene]hydroxylamine;chloride

vibrant-m
Empirical Formula (Hill Notation):
C7H9ClN2O
CAS Number:
Molecular Weight:
172.61

Products

Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Special Grade