Naphazoline nitrate
Synonym(s):
2-(1-Naphthylmethyl)imidazoline nitrate
Empirical Formula (Hill Notation):
C14H14N2 · HNO3
CAS Number:
Molecular Weight:
273.29
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing