
Myricetin 3-galactoside

Synonym(s):
Myricetin 3-O-β-D-Galactopyranoside
Empirical Formula (Hill Notation):
C21H20O13
CAS Number:
Molecular Weight:
480.38
Products
Formula Weight
Manufacturer
Product Line
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing