![Monolinuron certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/291/448/4740ce84-7f5f-43f2-a613-474c30ce75c9/640/4740ce84-7f5f-43f2-a613-474c30ce75c9.png)
Monolinuron
![Monolinuron certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/291/448/4740ce84-7f5f-43f2-a613-474c30ce75c9/640/4740ce84-7f5f-43f2-a613-474c30ce75c9.png)
Empirical Formula (Hill Notation):
C9H11ClN2O2
CAS Number:
Molecular Weight:
214.65
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing