![Monoclonal Anti-Protein A−Biotin antibody produced in mouse clone SPA-27, purified immunoglobulin, buffered aqueous solution](/deepweb/assets/sigmaaldrich/product/images/232/102/66c74e98-6914-4671-b346-7ac907d293b0/640/66c74e98-6914-4671-b346-7ac907d293b0.jpg)
Monoclonal Anti-Protein A−Biotin antibody produced in mouse
![Monoclonal Anti-Protein A−Biotin antibody produced in mouse clone SPA-27, purified immunoglobulin, buffered aqueous solution](/deepweb/assets/sigmaaldrich/product/images/232/102/66c74e98-6914-4671-b346-7ac907d293b0/640/66c74e98-6914-4671-b346-7ac907d293b0.jpg)
Synonym(s):
Monoclonal Anti-Protein A
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing