Monoclonal Anti-Pan Cadherin antibody produced in mouse
Synonym(s):
Monoclonal Anti-Pan Cadherin
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Antibody Form
Research Area
Conjugate
Application
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing