![Mirabegron CRS, European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/196/341/7473061f-3d2a-4295-9fa9-fbbfac296651/640/7473061f-3d2a-4295-9fa9-fbbfac296651.png)
Mirabegron
![Mirabegron CRS, European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/196/341/7473061f-3d2a-4295-9fa9-fbbfac296651/640/7473061f-3d2a-4295-9fa9-fbbfac296651.png)
Synonym(s):
2-(2-Amino-1,3-thiazol-4-yl)-N-[4-[2-[((2R)-2-hydroxy-2-phenylethyl)amino]ethyl]phenyl]acetamide, 2-Amino-N-[4-[2-[[(2R)-2-hydroxy-2-phenylethyl]amino]ethyl]phenyl]-4-thiazoleacetamide, YM 178, YM178(R)-Mirabegron, Mirabegron
Empirical Formula (Hill Notation):
C21H24N4O2S
CAS Number:
Molecular Weight:
396.51
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Research Area
Application
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing