Skip to Content
Merck
Metoxuron PESTANAL®, analytical standard

Metoxuron

Metoxuron PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C10H13ClN2O2
CAS Number:
Molecular Weight:
228.68
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique