Skip to Content
Merck
Methyl-trioctylammonium bis(trifluoromethylsulfonyl)imide for electrochemistry, ≥99% (NMR)

Methyl-trioctylammonium bis(trifluoromethylsulfonyl)imide

Methyl-trioctylammonium bis(trifluoromethylsulfonyl)imide for electrochemistry, ≥99% (NMR)
Synonym(s):
Methyltri-n-octylammonium bis(trifluoromethanesulfonyl)imide, Trioctylmethylammonium bis(trifluoromethylsulfonyl)imide
Empirical Formula (Hill Notation):
C27H54F6N2O4S2
CAS Number:
Molecular Weight:
648.85

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1