Skip to Content
Merck
Methyl myristate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Methyl myristate

Methyl myristate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Methyl tetradecanoate, Myristic acid methyl ester, Methyl myristate
Linear Formula:
CH3(CH2)12COOCH3
CAS Number:
Molecular Weight:
242.40
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Organoleptic Class
Package Size
API Family
Reference Material Type
Special Grade
Technique