Skip to Content
Merck
Methyl β-naphthyl ketone ≥99%, FCC, FG

Methyl β-naphthyl ketone

Methyl β-naphthyl ketone ≥99%, FCC, FG
Synonym(s):
2-Acetonaphthone, 2′-Acetonaphthone, 2-Acetylnaphthalene, Methyl 2-naphthyl ketone
Linear Formula:
C10H7COCH3
CAS Number:
Molecular Weight:
170.21
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing