![Methyl 3-[[(2RS)-2-(butylamino)propanoyl]amino]-4-methylthiophene-2- carboxylate hydrochloride certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/893/391/e397da5f-4e7a-496d-9684-713567208d0d/640/e397da5f-4e7a-496d-9684-713567208d0d.png)
Methyl 3-[[(2RS)-2-(butylamino)propanoyl]amino]-4-methylthiophene-2- carboxylate hydrochloride
![Methyl 3-[[(2RS)-2-(butylamino)propanoyl]amino]-4-methylthiophene-2- carboxylate hydrochloride certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/893/391/e397da5f-4e7a-496d-9684-713567208d0d/640/e397da5f-4e7a-496d-9684-713567208d0d.png)
Synonym(s):
Butylarticaine hydrochloride
Empirical Formula (Hill Notation):
C14H22N2O3S · HCl
CAS Number:
Molecular Weight:
334.86
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1