
Memantine Related Compound E

Synonym(s):
N-(3,5-Dimethyladamantan-1-yl)formamide, 1-Formylamino-3,5-dimethyladamantane, N-(3,5-Dimethyltricyclo[3.3.1.13,7]dec-1-yl)formamide
Empirical Formula (Hill Notation):
C13H21NO
CAS Number:
Molecular Weight:
207.31
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2