Meloxicam Related Compound D
Synonym(s):
4-Methoxy-2-methyl-N-(5-methyl-2-thiazolyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-dioxide
Empirical Formula (Hill Notation):
C15H15N3O4S2
Molecular Weight:
365.43
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing