![Luperox® DHD-9, 2-Butanone peroxide solution ~32 wt. % in phthalate-free plasticizer mixture](/deepweb/assets/sigmaaldrich/product/structures/349/187/925e4d0f-e56e-4355-a779-02c7416e9231/640/925e4d0f-e56e-4355-a779-02c7416e9231.png)
Luperox® DHD-9, 2-Butanone peroxide solution
![Luperox® DHD-9, 2-Butanone peroxide solution ~32 wt. % in phthalate-free plasticizer mixture](/deepweb/assets/sigmaaldrich/product/structures/349/187/925e4d0f-e56e-4355-a779-02c7416e9231/640/925e4d0f-e56e-4355-a779-02c7416e9231.png)
Synonym(s):
2-Butanone peroxide solution, Methyl ethyl ketone peroxide solution
Linear Formula:
C2H5C(OOH)(CH3)OOC(OOH)(CH3)C2H5
CAS Number:
Molecular Weight:
210.22
Products
Brand
Formula Weight
Quality Segment
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing