
L-Tyrosine-(phenyl-d4)

Synonym(s):
L-4-Hydroxyphenyl-2,3,5,6-d4-alanine
Linear Formula:
4-(HO)C6D4CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
185.21
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Optical Isomer
Showing 1-2 of 2
Compare
Product Number
Description
Pricing