Skip to Content
Merck
L-Leucine-5,5,5-d3 endotoxin tested, 99 atom % D

L-Leucine-5,5,5-d3

L-Leucine-5,5,5-d3 endotoxin tested, 99 atom % D
Linear Formula:
CH3CH(CD3)CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
134.19

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing