Liquiritin
Synonym(s):
4′,7-Dihydroxyflavanone 4′-glucoside, Liquiritoside
Empirical Formula (Hill Notation):
C21H22O9
CAS Number:
Molecular Weight:
418.39
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing