![Linezolid Related Compound C pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/738/721/63a79eef-1d01-43d1-875a-a26bbbf43a2f/640/63a79eef-1d01-43d1-875a-a26bbbf43a2f.jpg)
Linezolid Related Compound C
![Linezolid Related Compound C pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/738/721/63a79eef-1d01-43d1-875a-a26bbbf43a2f/640/63a79eef-1d01-43d1-875a-a26bbbf43a2f.jpg)
Synonym(s):
(5S)-5-(Aminomethyl)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-2-oxazolidinone, (S)-5-(Aminomethyl)-3-(3-fluoro-4-morpholinophenyl)oxazolidin-2-one
Empirical Formula (Hill Notation):
C14H18FN3O3
CAS Number:
Molecular Weight:
295.31
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2